Molybdenum pentachloride ≥99.6% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

11832.06 11832.18 11832.30 11832.A1
11832.06EA 37.5 EUR
11832.06 11832.18 11832.30 11832.A1
Molybdenum pentachloride ≥99.6% (metals basis)
Molybdenum pentachloride

Purity: 99.6% CAS No.: 10241-05-1 Molecular Formula: MoCl5 Formula Weight: 273.24 Physical Form: Crystalline Sensitivity: Moisture Sensitive Appearance: Black Odour: Pungent Melting Point: 194° UN No.: 2508 MDL No.: MFCD00003467

Formula: Cl₅Mo
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 194 °C
Density: 2.928 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003467
CAS Number: 10241-05-1
EINECS: 233-575-3
UN: 2508
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR