Sharp disposal containers for hazardous waste, Sharpsafe®
Supplier: Hospidex
SHA-41005410 SHA-41005430 SHA-41105430 SHA-41172430 SHA-41152430 SHA-41105410 SHA-41172410 SHA-41152410 SHA-41202410 SHA-41802410 SHA-41005420 SHA-41105420 SHA-41172420 SHA-41152420 SHA-41802420 SHA-41005440 SHA-41105440 SHA-41172440 SHA-41152440 SHA-41802440 SHA-41455430 SHA-41405430 SHA-41602430 SHA-41502430 SHA-41902430 SHA-41202411 SHA-41202421 SHA-41202441 SHA-41802411 SHA-41802421 SHA-41802441
119-0181EA 369 EUR
119-0181 119-0078 119-0075 119-0179 119-0186 119-0183 119-0187 HUARSHA-41152410 119-0189 119-0193 119-0182 119-0184 119-0188 119-0185 HUARSHA-41802420 HUARSHA-41005440 HUARSHA-41105440 HUARSHA-41172440 HUARSHA-41152440 HUARSHA-41802440 119-0191 119-0180 119-0192 HUARSHA-41502430 HUARSHA-41902430 HUARSHA-41202411 HUARSHA-41202421 HUARSHA-41202441 HUARSHA-41802411 HUARSHA-41802421 HUARSHA-41802441
Sharp disposal containers for hazardous waste, Sharpsafe®
Bins Sharps Containers
PP, yellow. Disposable plastic sharps container.
Orange lids – Sharps for incineration or alternative treatment. Marked 'Fully Discharged Sharps' the inscription and 'tick' indicate this coloured lid container is for use with sharps not containing prescription only medicines or 'POM's.
Yellow lids – Infectious sharps for incineration only. Marked with 'Medicinal Sharps', the inscription and 'tick' indicates this coloured lid container will be for general use, including sharps containing medicinal residue.
Purple lids – Marked with 'Cyto Sharps', the inscription and 'tick' indicateMore Product Information