Bench cans for solvents
Supplier: JUSTRITE MANUFACTURING
10175 10295 10375
119-0262EA 89.9 EUR
119-0262 119-0266 119-0269
Bench cans for solvents
Safety Cans
These bench cans for solvent safety have plated steel dasher which work as an effective flame arrester to reduce the risk of fire flashback. Hinged cover minimises evaporation losses and protected rim reduces the possibility of hand injury.
  • Sturdy 24-gauge steel body construction with tough powder-coat finish
  • Compliance with FM and TUV approved