Safety Transport Boxes
Supplier: STOROPACK
50001924 50001931 50001932 50001933 50001934 50001939 50001938
119-1550EA 208 EUR
119-1550 119-1551 119-1552 119-1553 119-1554 STOR50001939 STOR50001938
Safety Transport Boxes
Boxes Transport Boxes
EPS, white.
For bottles with dangerous content.
Certifications: BAM 8201/4H1