Sulphuric acid ∼96% in water for analysis

Supplier: Thermo Fisher Scientific
Danger

124640010 124640025 124645000 124640011 124640026 124640100 124645001
124640010.EA 45.9 EUR
124640010. 124640025. 124645000. 124640011. 124640026. 124640100. 124645001.
Sulphuric acid ∼96% in water for analysis
Sulphuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 10 °C
Density: 1.83 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Residue after ignition ≤0.0005 %
Titration with NaOH 95 to 97 %
Chloride (Cl) ≤0.5 ppm
Arsenic (As) ≤0.01 ppm
Iron (Fe) ≤0.5 ppm
Heavy metals (as Pb) ≤2 ppm
KMnO4-reducing substance ≤0.0008 % (as SO2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR