Potassium hexaiodoplatinate(IV) (≥18,2% Pt)

Supplier: Thermo Fisher Scientific
Warning

12642.03 12642.06
12642.03EA 126 EUR
12642.03 12642.06
Potassium hexaiodoplatinate(IV) (≥18,2% Pt)
Potassium hexaiodoplatinate (IV)

CAS No.: 16905-14-9 Molecular Formula: K2PtI6 Formula Weight: 1034.72 Physical Form: Crystalline MDL No.: MFCD00049660

Formula: I₆K₂Pt
Storage Temperature: Ambient
MDL Number: MFCD00049660
CAS Number: 16905-14-9
EINECS: 240-954-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR