Aluminium extension handles
Supplier: MICRONOVA
SSA-2
129-0282EA 94.9 EUR
129-0282
Aluminium extension handles
Mops
An ultra-lightweight, aluminum handle featuring three telescoping sections that extend to the desired length. Ideal for reaching high walls and ceilings, inside and around fixtures, and a range of cleanroom support applications.
  • Three telescoping sections for custom lengths
  • Ideal for extended reach in a range of cleanroom support applications
  • Available with extension from 6' to 18' (2 to 5,5 m)
  • Fits most mop frames and cleaning tools with threaded attachment sites
The Micronova Aluminium Extendible Mop Handles feature an ACME threaded end which fitsMore Product Information