Slim Jim® Vented Waste Bins, 60 and 87 L
Supplier: Rubbermaid Commercial Products
FG354007BLUE FG354060GRAY FG354007GRN FG354060BEIG FG354060BLA 1956188 1955959 1971259 1956187 1956181 1955960 1956189 1971257 1971258
129-0330EA 152 EUR
129-0330 129-0331 129-0428 129-0429 129-0430 129-2091 129-2095 129-2027 129-2092 129-2093 129-2094 RUBB1956189 129-3957 RUBB1971258
Slim Jim® Vented Waste Bins, 60 and 87 L
Bins Waste Bins
Enable easy waste management and reduce worker strain in any facility.
  • Sleek design with small footprint maximizes space to fit in the tightest areas
  • Venting channels make removing liners up to 80% easier, preventing unnecessary injury and improving productivity
  • Bag cinches are integrated into the side handles, to make securing liners simple
  • Robust molded handles, situated on the side and base of the container, provide improved control while lifting and emptying
  • The rib-strengthened rim resists crushing and prevents the containers from jamming when stacked