Cleaning tools, EasyReach™
Supplier: CONTEC
2644 MEQT0001 2665LF 2685LF MEQT0021 MEQT0022 MEQT0002SZ 2685P PSME0001-BPR
129-0204EA 378 EUR
129-0204 129-0574 CONT2665LF CONT2685LF 129-1655 129-1656 129-2470 CONT2685P 115-2814
Cleaning tools, EasyReach™
Mops
EasyReach™ is recommended for effective cleaning and disinfection of surfaces of separative devices such as: Isolators, restricted access barrier systems (RABS), laminar airflow hoods, bio safety cabinets, glove boxes, incubators and clean air hoods.
  • Specially designed tab on the cleaning pad enables quick and easy fitting to the frame with gloved hands
  • Single piece stainless steel tool is easily passed through transfer hatches, requires no assembly and is maintenance-free
  • Stainless steel tool is autoclavable for reuse in sterile environments
  • Versatile low