Steam sterilisation tubing, continuous form, Tyvek® 1073B PE
Supplier: KEYSTONE CLEANROOM PRODUCTS
BBTMGTVK04 BBTMGTVK06 BBTMGTVK09 BBTMGTVK12 BBTMGTVK14 BBTMGTVK18 BBTMGTVK24
129-3361EA 787 EUR
129-3361 129-3362 129-3363 129-3364 129-3365 129-3366 129-3368
Steam sterilisation tubing, continuous form, Tyvek® 1073B PE
Bags Sterilisation Pouches
Improve the prep time, performance, microbial barrier and cleanliness of your autoclave peel pouches.
  • Manufactured and packaged in a controlled cleanroom environment
  • Sizing flexibility and hydrophobic
  • Plastic core (no corrugated material)
  • Unroll and cut to desired length, heat seal both ends
  • Autoclavable at 121 to 125 °C
  • Peels open cleanly, lint-free
  • Tear and puncture resistant
A significant upgrade over paper based steam sterilisation tubing . These Tyvek® peel pouches will not rip or puncture like paper based autoclave pouches routinely do. TheMore Product Information