Blue Heavy-Duty Autoclavable SMS Muff Covers
Supplier: KEYSTONE CLEANROOM PRODUCTS
MFKCRP300-05 MFKCRP300-07 MFKCRP300-16 MFKCRP300-18 MFKCRP300-03 MFKCRP300-09 MFKCRP300-12 MFKCRP300-04 MFKCRP300-06 MFKCRP300-08
129-4011EA 1810 EUR
129-4011 129-4007 129-4021 129-4024 129-4014 129-4008 129-4012 KACCMFKCRP300-04 KACCMFKCRP300-06 KACCMFKCRP300-08
Blue Heavy-Duty Autoclavable SMS Muff Covers
Covers
This 50 g/m² SMS autoclavable muff cover provides an excellent microbial barrier that is ideal for pharmaceutical production applications.
  • Eliminates autoclave tape residue
  • Hydrophobic, tear-resistant, and puncture-resistant
  • Creates secondary level of protection during aseptic presentation and setup of equipment
  • Designed for autoclave cycles (121 to 135 °C)
  • Manufactured and packed in a certified ISO 8 cleanroom
  • Used in conjunction with a secondary autoclave bag, it creates an effective wrapping system
  • Made in USA
Eliminate the need for cutting sheetsMore Product Information