Gold for bright brushing

Supplier: Thermo Fisher Scientific
Danger

12943.04
12943.04EA 106 EUR
12943.04
Gold for bright brushing
Gold

Bright brushing gold.


Produces a bright continuous gold film on glass and ceramics when properly fired. Firing instructions included.

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 156 °C
Melting Pt: 1063
Flash Pt: 33 °C (91 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
UN: 1993
ADR: 3 III
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR