Vacuum controller, complete, VACUU·SELECT
Supplier: VACUUBRAND
20700060 20700070 20700080
131-0005EA 2455 EUR
131-0005 131-0006 131-0007
Vacuum controller, complete, VACUU·SELECT
Pumps Vacuum Controllers
The VACUU·SELECT complete controller is ready for use with existing vacuum sources like pumps or vacuum networks.
  • Pre-defined vacuum applications and automatic boiling point detection saves time
  • Simple set-up - connect VACUU·SELECT in-line between pump and vacuum application
  • Integrated vent valve allows inert gas connection
  • Integrated check valve minimises interaction with other applications on the same vacuum supply
The intuitive touch screen display offers pre-defined tools for all common vacuum processes. In evaporative processes, the first vapourMore Product Information