Chemistry diaphragm pumps, VARIO® select
Supplier: VACUUBRAND
20732450 20732451 20736550 20736551
131-0018EA 4695 EUR
131-0018 131-0019 131-0035 131-0036
Chemistry diaphragm pumps, VARIO® select
Pumps Vacuum Diaphragm Pumps
The VACUU·SELECT controller simplifies your work and allows VARIO select chemistry diaphragm pumps to deliver unparalleled precision. The pumps run whisper quiet and respond to demand in real time by matching pumping speed to process conditions. The intuitive VACUU·SELECT controller includes predefined applications for all common vacuum processes.
  • VACUU·SELECT vacuum controller with graphical user interface and predefined applications simplifies work in the laboratory
  • Automatic boiling point detection and adjustment of the vacuum for short process times