Cleanroom mops, PPSC Micro TEC
Supplier: PFENNIG REINIGUNGSTECHNIK
2700531
141-1393EA 22.8 EUR
141-1393
Cleanroom mops, PPSC Micro TEC
Mops
Mops provides top cleaning performance. Made up of 100% PES super microfiber with soft-brush.
  • Ready-to-use pre-washed
  • Backfilling with cover sheet, external trimming TEC
  • Pocket of harness extremely tear-proof
Germ reduction of bacteria, yeast fungus and mould of over 99 per cent will be reached by using water (Testing method DGMH 2001. standard method 14).
Certifications: Certified according to the requirements of the Nordic Ecolabel.