Zinc standard dissolved in oil, 1000 µg/g Zn in mineral oil for wear metal analysis, Specpure®

Supplier: Thermo Fisher Scientific

14200.1Y 14200.18
14200.1YEA 0 EUR
14200.1Y 14200.18
Zinc standard dissolved in oil, 1000 µg/g Zn in mineral oil for wear metal analysis, Specpure®
Standards Cationic Standards Zinc Standards

Physical Form: Liquid
MDL No.: MFCD00011291

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No. MFCD00011291

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR