Potassium standard dissolved in oil, 1000 µg/g K in mineral oil for wear metal analysis, Specpure®

Supplier: Thermo Fisher Scientific

14216.18
14216.18EA 104 EUR
14216.18
Potassium standard dissolved in oil, 1000 µg/g K in mineral oil for wear metal analysis, Specpure®
Standards Cationic Standards Potassium Standards

Physical form: liquid
MDL No.: MFCD00133776

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR