Ammonium bromide ≥99.0% ACS

Supplier: Thermo Fisher Scientific
Warning

14257.14 14257.22 14257.36 14257.A3
14257.14EA 30.3 EUR
14257.14 14257.22 14257.36 14257.A3
Ammonium bromide ≥99.0% ACS
Ammonium bromide

Purity: 99.0% min CAS No.: 12124-97-9 Molecular Formula: NH4Br Formula Weight: 97.97 Physical Form: Granular Density: 2.429 Appearance: White Melting Point: 452° subl. Solubility: Freely soluble in water, methanol, ethanol, and acetone. Slightly soluble in ether. Practically insoluble in ethyl acetate Merck Reference: 14,505 MDL No.: MFCD00011419


Corrosion inhibitors, manufacturing of photographic films, plates, paper, lithography, fireproofing of woods

Formula: H₄BrN
Melting Pt: 452 °C (subl.)
Density: 2.429
Storage Temperature: Ambient
MDL Number: MFCD00011419
CAS Number: 12124-97-9
EINECS: 235-183-8
Merck Index: 14,505

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR