Neodymium trichloride, ultra dry ≥99.99% (REO, rare earth oxide basis)

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Neodymium (III) chloride

ampouled under argon

14282.06 14282.14
14282.06EA 301 EUR
14282.06 14282.14
Neodymium trichloride, ultra dry ≥99.99% (REO, rare earth oxide basis)
Neodymium trichloride

Purity: 99.99% CAS No.: 10024-93-8 Molecular Formula: NdCl3 Formula Weight: 250.60 Physical Form: -20 Mesh Powder, Ampouled under argon Density: 4.134 Melting Point: 784° Merck Reference: 14,6450 MDL No.: MFCD00011132

Formula: Cl₃Nd
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 784 °C
Density: 4.134
Storage Temperature: Ambient
MDL Number: MFCD00011132
CAS Number: 10024-93-8
EINECS: 233-031-5
Merck Index: 14,6450

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR