Potassium standard solution, 10000 mg/L K in 5% nitric acid (from KNO₃), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific

14379.AD 14379.AE 14379.AP
14379.ADEA 108 EUR
14379.AD 14379.AE 14379.AP
Potassium standard solution, 10000 mg/L K in 5% nitric acid (from KNO₃), standard for ICP, Specpure®
Standards Cationic Standards Potassium Standards

Molecular Formula: KNO₃ in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00133776


Certifications: Traceable to NIST

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No. MFCD00133776

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR