Calcium sulphate dihydrate 98.0-102.0%, powder, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor

1452-01EA 0 EUR
1452-01
Calcium sulphate dihydrate 98.0-102.0%, powder, BAKER ANALYZED® ACS, J.T.Baker®
Calcium sulphate dihydrate

Powder. Lot analysis on label.

Formula: CaSO₄·2H₂O
Density: 2.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149625
CAS Number: 10101-41-4
EINECS: 231-900-3
Merck Index: 13,01711

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements 98.0 - 102.0 %
For Laboratory,Research,or Manufacturing Use 98.0 - 102.0 %
Assay (CaSO₄·2H₂O) (by EDTA titrn) 98.0 - 102.0 %
Carbonate (CO₃) Passes Test
Chloride (Cl) ≤0.002 %
Heavy Metals (as Pb) ≤0.002 %
Insoluble in Dilute HCl ≤0.02 %
Trace Impurities - Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.02 %
Nitrate (NO₃) Passes Test
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.02 %
Strontium (Sr) ≤0.05 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR