Gold ≥99.999% (metals basis), wire, Premion®, Ø 1.0 mm (0.04 in)

Supplier: Thermo Fisher Scientific

14726.BP 14726.BS 14726.BW
14726.BPEA 104 EUR
14726.BP 14726.BS 14726.BW
Gold ≥99.999% (metals basis), wire, Premion®, Ø 1.0 mm (0.04 in)
Gold

Purity: 99.999% CAS No.: 7440-57-5 Physical Form: ≈15.15g/m Odour: Odourless MDL No.: MFCD00003436

Formula: Au
Boiling Pt: 2800 °C (1013 hPa)
Melting Pt: 1063 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR