Detergents, liquid, for automated cleaning of glassware, neodisher® FA
Supplier: Dr. Weigert
410126 410133 410130
148-3138EA 310 EUR
148-3138 148-3142 148-0162
Detergents, liquid, for automated cleaning of glassware, neodisher® FA
Cleaning Agents Detergents and Surface Cleansers
neodisher® FA.
  • Alkaline detergent for the automated reprocessing of thermostable and thermolabile instruments
  • Components: 15 to 30% phosphates
  • Dosage: 2 to 4 ml/L
Components: 15 - 30% phosphates, Disodium and potasium metasilicate.
Dosage: 2 - 4 ml/l
Alkaline liquid concentrate for universal applications Free of phosphates, surfactants and oxidising agents.
Components: 5 - 15% polycarboxylates, NTA (nitrilotriacetic acid).
Caution: Strong exothermic reaction with acids. Corrodes aluminium.More Product Information