Gas detection tube for sulphuric acid
Supplier: Dräger
6728781
151-0159EA 126 EUR
151-0159
Gas detection tube for sulphuric acid
Gas Tubes
The data below is representative for use with an Accuro® pump, a hand operated bellows pump.
  • Soluble sulfates and other aerosol forming acids indicated but with different sensitivities
  • Impossible to measure sulfuric acid in the presence of these substances
  • Sulfur trioxide in the gaseous form not indicated (but in the presence of atmospheric humidity this forms sulfuric acid aerosols, which are indicated)
One stroke is equivalent to a volume of 100 ±5 ml of gas passing through the tube.
Operating conditions: 5 to 40 °C ; Up to 15 mg/L H₂O absoluteMore Product Information