Air flow indicator
Supplier: Dräger
CH00216
151-0187EA 111 EUR
151-0187
Air flow indicator
Leak Detection Systems Gas Leak Detection Systems
This air flow indicator for special applications can be used in all operations relating to draught detection.
Delivery information: Supplied with an aspirator bulb, ten air flow test tubes and two sealing caps.