1-Octanesulphonic acid sodium salt 98, HiPerSolv CHROMANORM® HPLC

Supplier: VWR Chemicals
Warning

Synonyms: Sodium octane-1-sulphonate

152802TEA 237 EUR
152802T 152803U
1-Octanesulphonic acid sodium salt 98, HiPerSolv CHROMANORM® HPLC
1-Octanesulphonic acid sodium salt
Formula: C₈H₁₇NaO₃S
MW: 216.28 g/mol
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 5324-84-5
EINECS: 226-195-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1244cb11db07775dec2e193823e40080024b7036&vid=32d95cd9e321818e2f94c8261c4b4e6180ee0fc7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated on dried substance) ≥ 98,0 %
Appearance White powder
IR Spectrum Passes test
Water ≤ 2,0 %
Absorbance (200 nm) (0.25 mol/l) ≤ 0,1
Absorbance (210 nm) (0.25 mol/l) ≤ 0,05
Absorbance (220 nm) (0.25 mol/l) ≤ 0,03
Absorbance (230 nm) (0.25 mol/l) ≤ 0,02
Absorbance (240 nm) (0.25 mol/l) ≤ 0,01
Absorbance (250 nm) (0.25 mol/l) ≤ 0,01
Absorbance (260 nm) (0.25 mol/l) ≤ 0,01


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR