Affinity chromatography columns, HiTrap™ Heparin HP
Supplier: Cytiva
17-0406-01 17-0407-01 17-0407-03
17-0406-01EA 304 EUR
17-0406-01 17-0407-01 17-0407-03
Affinity chromatography columns, HiTrap™ Heparin HP
Chromatography Columns
HiTrap Heparin HP columns are designed for high-resolution purification of DNA-binding proteins, coagulation factors, and other plasma proteins.
  • Suitable for purification of antithrombin III, coagulation factors, and other plasma proteins
  • Useful for screening and optimisation before scaling up to bulk resins such as Heparin 6 Fast Flow
  • High-resolution purification and elution in narrow peaks, minimizing the need for further concentration steps
  • Prepacked with Heparin Sepharose High Performance resin
  • HiTrap format compatible with syringes, pumps, and