Water purification systems, Milli-Q® IQ 7003/05/10/15
Supplier: Merck
ZIQ7003T0C ZIQ7005T0C ZIQ7010T0C ZIQ7015T0C
171-1378EA 19720 EUR
171-1378 171-1379 171-1380 171-1381
Water purification systems, Milli-Q® IQ 7003/05/10/15
Water Purification Systems Pure and Ultra Pure Water Systems
An integrated ultrapure and pure water system that is intelligent, easy to use and environmentally friendly.
  • Delivers both ultrapure and pure water from up to four POD dispensers
  • Four pure water production flow rates (3, 5, 10 or 15 L/h)
  • Three tanks sizes (25, 50 or 100 L)
  • Space-saving and clutter-free installation options, on the wall, under the bench, or both
  • Large selection of application POD-Paks, to meet your every application need
  • Tailored service plans and a range of additional support options
The Milli-Q® IQ 7003/7005/7010/7015 system is aMore Product Information