Pure water systems, Milli-Q® IX 7003/05/10/15
Supplier: Merck
ZIX7003P0C ZIX7015P0C ZIX7003T0C ZIX7005P0C ZIX7005T0C ZIX7010P0C ZIX7010T0C ZIX7015T0C
172-0055EA 12120 EUR
172-0055 172-0061 172-0056 172-0057 172-0058 172-0059 172-0060 172-0062
Pure water systems, Milli-Q® IX 7003/05/10/15
Water Purification Systems Pure and Ultra Pure Water Systems
Reliable water purification for results reproducibility and lab productivity.
  • Constant-quality Elix® pure water
  • Intuitive E-POD® dispenser (optional)
  • Touchscreen interface
  • New pure water storage solution
  • Elix® EDI technology lowers running costs
  • Mercury-free UV lamps
The Milli-Q IX pure water system provides a reliable source of high quality type 2 pure water to feed your lab's needs: From general laboratory uses, such as instrument feed or buffer preparation, to sensitive applications, such as microbiology culture media or staining solutions.