MXPC diaphragm pumps, Welch Vacuum
Supplier: Welch by Gardner Denver
117047 117057
181-0644EA 3100 EUR
181-0644 181-0645
MXPC diaphragm pumps, Welch Vacuum
Pumps Vacuum Pumps
The MXPC SERIES comes from the latest generation of Welch MPCpro diaphragm pumps to provide improved performance, particularly in the application critical range. The ultimate vacuum of 5 or 1,5 mbar with significant design improvements to directly influence the process flow and the repeatability. The new Single Piece Diaphragm (SPD) design provides increased chemical resistance and easier servicing. These models are ideal for applications where a high pumping speed, fast pump down time and deep ultimate vacuum are required, such as vacuum ovens, rotary evaporators and vacuum concentrators.