Vacuum hand pumps, Nalgene®
Supplier: Thermo Fisher Scientific
6131-0010 6131-0020 6132-0010 6132-0020 6133-0010 6132-2000 6132-3000
181-0434EA 185 EUR
181-0434 181-9907 181-9902 181-9903 181-9904 181-0473 181-0474
Vacuum hand pumps, Nalgene®
Pumps Vacuum Hand Pumps
With these sturdy pumps, a partial vacuum of 86 mbar can be reached very quickly and easily. The vacuum can be controlled with just the index finger. Vacuum is set via a lever on the pump. The pumps reach full pressure with just two priming strokes. Removable cover on exhaust port and trigger to release vacuum, so the pump does not need to be disconnected from the partial vacuum line for venting. Hose connection for standard hoses with 6,4 mm internal diameter. Complete with 60 cm transparent plastic tube.
Vacuum hand pumps without vacuum gauge - PVC housing
Suction volume 36 ml per strokeMore Product Information