Methyl violet 2B, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Danger

Synonyms: Basic Violet 1, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]methylammonium chloride

M-Clarity™ quality level = MQ100

198099-100G 198099-25G
198099-100GEA 230 EUR
198099-100G 198099-25G
Methyl violet 2B, Sigma-Aldrich®
Methyl Violet 2B
Formula: C₂₄H₂₈ClN₃
MW: 393.96 g/mol
Melting Pt: 137
Storage Temperature: Ambient
MDL Number: MFCD00012568
CAS Number: 8004-87-3
UN: 3077
ADR: 9 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2cbb5eadf0a61c4c5aa95151b8cb7ce8ca23557d&vid=6c464e6ab3c52e94db2297f4a6bcad662c2076d4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR