Sodium fluoride 87% ≥87% (w/v) in sodium hydroxide solution

Supplier: VWR Chemicals
Warning

19817DVEMPTYEA 0 EUR
19817DVEMPTY
Sodium fluoride 87% ≥87% (w/v) in sodium hydroxide solution
Sodium fluoride
Formula: NaF
MDL Number: MFCD00003524
CAS Number: 7681-49-4
EINECS: 231-667-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2807d129a60e4d136eb782386d584317338c4499&vid=02396e9a739aa8e35f0b20961710e28c6ec76be0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear Colourless Liquid
Sodium hydroxide (W/V) 0.0198 - 0.0242%
Fluoride (W/V) 1.300 - 1.365%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR