Acid red 1

Supplier: Thermo Fisher Scientific

198700050 198700250
198700050.EA 21.6 EUR
198700050. 198700250.
Acid red 1
Acid red 1
Formula: C₁₈H₁₅N₃O₈S₂
Storage Temperature: Ambient
MDL Number: MFCD00003954
CAS Number: 3734-67-6
EINECS: 223-098-9

Order Now

Specification Test Results

Appearance red to brown to purple powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Loss on drying ≤10 %
Specific absorbance A (1%/1cm) ≥250 lambda 503 - 509 nm, 0.02 g/l water


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR