Phenosafranine 80% (dye content), Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Warning

Synonyms: 3,7-Diamino-5-phenylphenazinium chloride

199648-1G
199648-1GEA 108 EUR
199648-1G
Phenosafranine 80% (dye content), Sigma-Aldrich®
Phenosafranine

Phenosafranin has been used for nuclear staining.


  • Phenosafranin (PSF) is a red dye released from cells with intact plasma membrane
  • PSF at lower concentration is used to stain mitochondria supravitally


It is also used a diagnostic assay manufacturing, hematology, histology.

Formula: C₁₈H₁₅ClN₄
Melting Pt: >300 °C
Storage Temperature: Room Temperature
CAS Number: 81-93-6
EINECS: 201-387-0
Merck Index: 12,07402
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3a8a6fac2d20e232b1d1ce1b1ad35b2ab72006d&vid=511626f64c7cf2a4641c5c792de099afe04f116c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Carbon ≥46.7%
Nitrogen ≥12.0%
Wavelength 517 - 523 nm
Extinction Coefficient ≥29000


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR