Bubble counters
Supplier: Lenz Laborglas GmbH & CO.KG
05596000
201-9100EA 33.4 EUR
201-9100
Bubble counters
Bottles Gas Washing Bottles
DURAN®, borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For monitoring gas flow in distillation apparatus.