Diisobutylaluminium hydride 1.0 M in heptane, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Di-i-butylaluminum hydride, DIBAL-H

201051000 201058000
201051000.EA 85.4 EUR
201051000. 201058000.
Diisobutylaluminium hydride 1.0 M in heptane, AcroSeal®
Diisobutylaluminium hydride
Formula: C₈H₁₉Al
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008928
CAS Number: 1191-15-7
EINECS: 214-729-9
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Clear solution
Strength 19.5 to 20.5 % (DIBAL-H, on Al basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR