Vydac 208MS C8, HPLC columns
Supplier: Avantor
208MS5210EA 857 EUR
208MS5210 208MS5215 208MS5415 208MS52 208MS54
Vydac 208MS C8, HPLC columns
Chromatography Columns
Vydac MS columns are a later development from Grace for the reversed phase HPLC separation of biomolecules. A proprietary surface treatment and bonding process give Vydac MS columns unique selectivity. A variety of reversed phases makes this product line suitable for the analysis of small peptides to large intact, undenatured proteins.
  • Five reversed phase chemistries
  • Excellent peak shape with little or no TFA
  • High protein recoveries make scale-up easy
Lower hydrophobicity which is better for larger biomolecules. Ideal for biomolecules with 5 to 10 kMore Product Information