Vydac 208TP C8, HPLC columns
Supplier: Avantor
554-7581EA 1210 EUR
554-7581 208TP510 208FSK510 208TP5210 208TP5215 208TP5415 208TP52 208TP1010 208TP1022 208GCC1210 208FSK1210 208TP101510 208TP152022 208GD104/N 208GD34/N 208GD52/N 208GD54/N 208GK104/N 208GK34/N 208GK52/N 208GK54/N 208GCC1010
Vydac 208TP C8, HPLC columns
Chromatography Columns
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations. The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
  • Long column lifetime and negligible phase leaching
  • Reliable protein purifications, scalable from analytical to preparative scale
  • Referenced in a large number of patents and publications
208TP C8 is less hydrophobic than C18TP and is recommended for the analysis of peptides, 10 – 20KMore Product Information