Ethanol 96% vol Ph. Eur.

Supplier: VWR Chemicals
Danger

Synonyms: Ethyl alcohol

20905.296EA 63.2 EUR
20905.296 20905.365 20905.467 20905.320
Ethanol 96% vol Ph. Eur.
Ethanol

Caution: WARNING : This alcohol is regulated. Please contact VWR. For duty paid orders in the UK please add DP to the product code.

Formula: C₂H₅OH
MW: 46.07 g/mol
Boiling Pt: 78.3 °C (1013 hPa)
Melting Pt: –117 °C
Density: 0.7895 g/cm³ (20 °C)
Flash Pt: 12 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3 II
REACH: 01-2119457610-43
Merck Index: 12,03806

Order Now

Specification Test Results

Assay (V/V) 95.1 - 96.9 %
Appearance Conforms (see CoA/CoS)
Identification (IR) Passes test
Appearance test Passes test
Acidity or alkalinity Passes test
Relative density 0.805 - 0.812
Absorbance (240 nm) Max. 0.4
Absorbance (250-260 nm) Max. 0.30
Absorbance (270-340 nm) Max. 0.10
The absorption curve is smooth Passes test
Methanol (V/V) Max. 200 ppm
Acetaldehyde + acetal (as C2H4O) (V/V) Max. 10 ppm
Benzene (V/V) Max. 2 ppm
Impurities (as 4-methylpentan-2-ol)(V/V) Max. 300 ppm
Disregard limit Max. 9 ppm
Residue on evaporation (W/V) Max. 25 ppm
Residual solvents Passes test
Bacterial endotoxins max 2.5 IU/ml

SPECIFICATIONS

Environmentally Preferable Safer Human and Environmental Health
Environmental certification EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR