Phosphate-buffered saline (PBS), Corning®
Supplier: Corning
21-040-CV 21-040-CM 21-040-CMX12
21-040-CVEA 57.07 EUR
21-040-CV 21-040-CM 21-040-CMX12
Phosphate-buffered saline (PBS), Corning®
Buffers Cell Culture Buffers
Phosphate buffered saline (PBS) is a buffer solution containing sodium chloride, sodium phosphate, and in some formulations potassium chloride and potassium phosphate
  • Low endotoxin - <0.25 EU/ml
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
PBS helps to maintain a constant pH and is isotonic and non-toxic to cells.