Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
0030120.086 0030120.159 0030120.167 0030120.175 0030120.183 0030120.191 0030121.694 0030123.328 0030121.589 0030123.611
211-2130EA 51.5 EUR
211-2130 211-2131 211-2132 211-2133 211-2134 211-2135 211-2139 211-2164 211-2161 525-1042
Safe-Lock Tubes, 1,5 ml - Microtubes
Tubes Microcentrifuge Tubes
Eppendorf® Safe-Lock 1,5 ml microtubes ensure secure sample storage. With a patented Safe-Lock design, these microtubes prevent accidental opening, safeguarding samples. Ideal for diverse applications, they offer reliability, durability, and ease of use in a 1,5 ml capacity.
  • Hinged Safe-Lock lid prevents inadvertent lid opening during incubation and storage
  • g-Safe® technology, depending on the Safe-Lock Tube variant, ensures outstanding centrifugation stability up to 30,000×g
  • g-Safe® technology prevents sample loss due to tube breakage, and provides