Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Supplier: EPPENDORF
0030121.023 0030121.112 0030121.120 0030121.139 0030121.147 0030121.155 0030121.570 0030121.708 0030123.301 0030123.603
211-2140EA 50.9 EUR
211-2140 211-2141 211-2142 211-2143 211-2144 211-2145 211-2162 211-2149 211-2163 525-1041
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Tubes Microcentrifuge Tubes
Eppendorf® Safe-Lock tubes 0,5 ml - Microtube ensures secure sample storage. With a unique locking mechanism, these tubes prevent accidental opening, safeguarding precious samples. Ideal for PCR, these tubes offer reliability and ease in a compact design.
  • Hinged Safe-Lock lid prevents unintended opening during incubation and storage
  • g-Safe® technology, dependent on the Safe-Lock tube variant (refer to the manual), ensures exceptional centrifugation stability up to 30000×g
  • g-Safe® technology prevents sample loss in the event of tube breakage, and provides