Biotube™ system
Supplier: Simport Scientific
T100-1B T100-1G T100-1R T100-1Y T100-2B T100-2G T100-2R T100-2Y T100-3B T100-3G T100-3R T100-3Y T100-4B T100-4G T100-4R T100-4Y T100-50G T100-50R T100-50Y T100-60B T100-60G T100-60R T100-60Y T100-20 T100-25 T100-20LST T100-26 T100-35 T100-28 T100-30 T100-40
212-9550EA 130 EUR
212-9550 SIMPT100-1G SIMPT100-1R SIMPT100-1Y 212-9553 SIMPT100-2G SIMPT100-2R SIMPT100-2Y 212-9554 SIMPT100-3G SIMPT100-3R SIMPT100-3Y 212-9555 SIMPT100-4G SIMPT100-4R SIMPT100-4Y SIMPT100-50G SIMPT100-50R SIMPT100-50Y 720-0499 SIMPT100-60G SIMPT100-60R SIMPT100-60Y 212-9551 212-9552 SIMPT100-20LST SIMPT100-26 SIMPT100-35 SIMPT100-28 SIMPT100-30 SIMPT100-40
Biotube™ system
Tubes Microtubes
PP tube and rack system with tubes held in a standard 8×12 configuration.
  • Alphanumeric coding and removeable 96-place ID card for accurate sample location
  • Transparent cover keyed to base to prevent misalignment
  • Certified RNase, DNase, pyrogen- and DNA-free
  • Conforms to SBS standards
  • Caps are made of PE and are not autoclavable
  • Racks and tubes can be used over a wide temperature range, from −80 to +121 °C
These tubes are ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting, pharmaceutical quality control, receptorMore Product Information