Beakers, low form
Supplier: KARTELL
1822 1824 1821 1823 1825 1826 1827 818 819
213-1632EA 5 EUR
213-1632 213-1634 213-1631 213-1633 213-1641 213-1636 213-1643 213-1644 213-1645
Beakers, low form
Beakers
PP, highly translucent.
  • Good chemical resistance
  • Resistant to heat up to 121 °C (for 20 minutes), prolonged use at 100 °C
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use. With spout and easy to read blue graduations.
ISO 7056
Certifications: ISO 7056.