VWR®, Erlenmeyer Flasks
Supplier: VWR Collection
214-1130EA 75.7 EUR
214-1130 214-1131 214-1132 214-1133 214-1134 214-1136 214-1137 214-1170 214-1171 214-1172 214-1174 214-1175 214-1173 214-0248 214-0471 214-0472
VWR®, Erlenmeyer Flasks
Flasks Erlenmeyer Flasks
Borosilicate 3.3 glass, clear.
  • Narrow or wide neck, with beaded rim
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
An economical alternative for a wide range of laboratory applications. Flasks have flared rim with markings for approximate volume graduations and a white labelling area.
ISO 1773 (narrow mouth)
DIN ISO 24450 (wide mouth)
Certifications: ISO 1773 (narrow neck), DIN ISO 24450 (wide neck).