Bottles, narrow neck, with standard ground stoppers, PYREX®
Supplier: SciLabware
1521/08D 1521/16D 1521/18D 1521/22D 1521/04D 1521/06D 1521/10D 1521/12D
215-0342EA 46.1 EUR
215-0342 215-0345 215-0346 215-0347 215-0340 215-0341 215-0343 BIBB1521/12D
Bottles, narrow neck, with standard ground stoppers, PYREX®
Bottles Glass Bottles
PYREX®, borosilicate glass, with standard ground joint and glass stopper.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Marked with volume and a white labelling field.
ISO4796-2
Certifications: ISO4796-2.