HPLC bottles with conical base GL 45, DURAN®
Supplier: DWK Life Sciences
218417305 218415407 218418601 218416309 218416909
215-4546EA 486 EUR
215-4546 215-4543 215-4547 215-4544 215-4545
HPLC bottles with conical base GL 45, DURAN®
Bottles HPLC Bottles
DURAN®, borosilicate glass 3.3, clear, with blue PP screw cap.
  • Central cavity can accommodate magnetic stir bars for improved mixing
  • Side vent or drain hole in the base for ultrasonic bath degassing
  • Durable white enamel volume graduations
  • Autoclavable at 121 °C or 134 °C
Applications include mobile phase reservoirs for HPLC, reagent supply to liquid handling systems. DURAN® reservoirs have been specially designed for the storage and delivery of high performance liquid chromatography mobile phases and solvents. Reservoirs have one cavity for the filterMore Product Information