Safety wash bottles, narrow mouth, printed, VITsafe™
Supplier: VITLAB
1332819 1332839 1332849 1332859 1332869 1332879 1332889 1332899 1332909 1332939 1332949 1332959 1332969 1332979 1331819 1331839 1331849 1331859 1331869 1431829 1432829 1432989 1333819_U 1333839 1333849 1333859 1333869 1433829
215-5716EA 9.9 EUR
215-5716 215-5714 215-5715 215-5700 215-5702 215-5708 215-5701 215-5709 215-5710 215-5717 215-5703 215-5704 215-5705 215-5706 215-1716 215-6873 215-6874 215-1717 215-6840 215-6841 215-6858 215-6864 215-6875 215-6876 215-6877 215-1718 215-6878 215-6865
Safety wash bottles, narrow mouth, printed, VITsafe™
Bottles Wash Bottles
LDPE, with VENT CAP threaded connection.
  • Colour coded contents with hazard symbols and descriptions of hazards as required by the Hazardous Substance Ordinance
  • Good chemical resistance
  • Good thermal resistance
VENT CAP works on the principle of gas/vapour permeability. The screw cap prevents any build-up of vapour pressure above the contents. This prevents the risk of solvent dripping or leaking.
Caution: Note: Washbottle 'Hexan' is not suitable for n-Hexane.