Nalgene™ Bottles, Wide Neck, with Screw Cap
Supplier: Thermo Fisher Scientific
2120-0005 2120-0010 2121-0005 2120-0005Z 2120-0010Z 2121-0010
215-7351EA 22.2 EUR
215-7351 215-7352 215-7361 NALG2120-0005Z NALG2120-0010Z 215-1281
Nalgene™ Bottles, Wide Neck, with Screw Cap
Bottles Plastic Bottles
HDPE, translucent with PP screw cap or PPCO, translucent with PP screw cap.
  • HDPE: Good chemical resistance, heat resistant up to 120 °C
  • PPCO: Very good chemical resistance, heat resistant up to 121 °C, autoclavable
  • Extra wide opening
Large volume multipurpose bottles for a wide range of uses. Not leakproof.
These high-density, polyethylene bottles are good for use in the freezer to −100 °C (−148 °F). Chemical-resistant HDPE is compatible with a wide variety of lab reagents.
Linerless cap closes the bottle without a liner that can wrinkle, causeMore Product Information